Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-3-Methylvaleric acid 97%

Supplier: AMBEED, INC
Danger

A844839-10G A844839-25G A844839-5G
77281-678EA 11 USD InStock
77281-678 77281-680 77281-682
(±)-3-Methylvaleric acid 97%
(±)-3-Methylvaleric acid
Precaución: for Research Use Only


Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 196
Melting Pt: -41
MDL Number: MFCD00002729
CAS Number: 105-43-1
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty