Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Dichloropyrimidine 98%

Supplier: AMBEED, INC
Danger

A845165-25G A845165-100G
77281-690EA 6 USD InStock
77281-690 77281-692 77618-366
4,6-Dichloropyrimidine 98%
4,6-Dichloropyrimidine
Precaución: for Research Use Only


Formula: C₄H₂Cl₂N₂
MDL Number: MFCD00006109
CAS Number: 1193-21-1
UN: 3263
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty