Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dimethylthiophene 98%

Supplier: AMBEED, INC
Danger

A846254-25G A846254-5G
77256-284EA 34 USD InStock
77256-284 77256-286
2,5-Dimethylthiophene 98%
2,5-Dimethylthiophene
Precaución: for Research Use Only


Formula: C₆H₈S
MW: 112.2 g/mol
Boiling Pt: 134
Melting Pt: -63
MDL Number: MFCD00005452
CAS Number: 638-02-8
UN: 1993
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty