Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

PFK-158 ≥98%

Supplier: AMBEED, INC
Warning

A848978-10MG A848978-1MG A848978-25MG A848978-5MG
77244-500EA 42 USD InStock
77244-500 77244-502 77244-504 77244-506
PFK-158 ≥98%
PFK-158
Precaución: for Research Use Only


Formula: C₁₈H₁₁F₃N₂O
MDL Number: MFCD28386154
CAS Number: 1462249-75-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty