Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dichlorobenzoyl chloride 98%

Supplier: AMBEED, INC
Danger

A850027-25G A850027-5G
77281-708EA 21 USD InStock
77281-708 77281-710
3,4-Dichlorobenzoyl chloride 98%
3,4-Dichlorobenzoyl chloride
Precaución: for Research Use Only


Formula: C₇H₃Cl₃O
MW: 209.46 g/mol
Boiling Pt: 242
Melting Pt: 34
MDL Number: MFCD00000672
CAS Number: 3024-72-4
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty