Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2S,3S)-1,4-dioxaspiro[4.5]decane-2,3-diylbis(diphenylmethanol) ≥98%, ee 99%

Supplier: AMBEED, INC
Warning

A851620-100MG A851620-1G A851620-250MG A851620-5G
77256-320EA 12 USD InStock
77256-320 77256-322 77256-324 77256-326
(2S,3S)-1,4-dioxaspiro[4.5]decane-2,3-diylbis(diphenylmethanol) ≥98%, ee 99%
(2S,3S)-1,4-dioxaspiro[4.5]decane-2,3-diylbis(diphenylmethanol)
Precaución: for Research Use Only


Formula: C₃₄H₃₄O₄
MDL Number: MFCD09033383
CAS Number: 123287-35-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty