Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Iodo-1,3-benzodioxole 98%

Supplier: AMBEED, INC
Warning

A852516-10G A852516-1G A852516-25G A852516-5G
77244-520EA 27 USD InStock
77244-520 77244-522 77244-524 77244-526
5-Iodo-1,3-benzodioxole 98%
5-Iodo-1,3-benzodioxole
Precaución: for Research Use Only


Formula: C₇H₅IO₂
MW: 248.0179 g/mol
MDL Number: MFCD00079766
CAS Number: 5876-51-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty