Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Diiodosalicylic acid 98%

Supplier: AMBEED, INC
Warning

A854718-100G A854718-25G A854718-500G A854718-5G A854718-1000G
77281-722EA 53 USD InStock
77281-722 77281-724 77281-726 77281-728 77749-662
3,5-Diiodosalicylic acid 98%
3,5-Diiodosalicylic acid
Precaución: for Research Use Only


Formula: C₇H₄I₂O₃
MDL Number: MFCD00002444
CAS Number: 133-91-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty