Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-2-Butene-1,4-diol 95%

Supplier: AMBEED, INC
Warning

A857011-100G A857011-10G A857011-1G A857011-250MG A857011-5G
77268-610EA 707 USD InStock
77268-610 77268-612 77268-614 77268-616 77268-618
trans-2-Butene-1,4-diol 95%
trans-2-Butene-1,4-diol
Precaución: for Research Use Only


Formula: C₄H₈O₂
MDL Number: MFCD00063207
CAS Number: 821-11-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty