Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxyacetamide 97%

Supplier: AMBEED, INC
Warning

A858149-1G A858149-250MG A858149-25G A858149-5G A858149-100G
77268-628EA 10 USD InStock
77268-628 77268-630 77268-632 77268-634 77749-665
2-Hydroxyacetamide 97%
2-Hydroxyacetamide


Formula: C₂H₅NO₂
MDL Number: MFCD00047895
CAS Number: 598-42-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty