Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Thienylacetyl chloride 96%

Supplier: AMBEED, INC
Danger

A863124-5G A863124-25G
77281-750EA 11 USD InStock
77281-750 77749-671
2-Thienylacetyl chloride 96%
2-Thienylacetyl chloride
Precaución: for Research Use Only


Formula: C₆H₅ClOS
MW: 160.62 g/mol
MDL Number: MFCD00005456
CAS Number: 39098-97-0
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty