Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bempedoic acid 98%

Supplier: AMBEED, INC
Warning

A869454-10MG A869454-1MG A869454-250MG A869454-25MG A869454-5MG A869454-50MG
77244-574EA 61 USD InStock
77244-574 77244-576 77244-578 77244-580 77244-582 77747-641
Bempedoic acid 98%
Bempedoic acid
Precaución: for Research Use Only


Formula: C₁₉H₃₆O₅
MW: 344.49 g/mol
MDL Number: MFCD18800820
CAS Number: 738606-46-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty