Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

TRC051384 98+%

Supplier: AMBEED, INC
Warning

A875310-10MG A875310-1MG A875310-5MG
77268-706EA 85 USD InStock
77268-706 77268-708 77268-710
TRC051384 98+%
TRC051384
Precaución: for Research Use Only


Formula: C₂₅H₃₁N₅O₄
MDL Number: MFCD28133369
CAS Number: 867164-40-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty