Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-(Nd-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl)-D-ornithine 97%

Supplier: AMBEED, INC
Warning

A876627-100MG A876627-1G A876627-250MG A876627-5G
77244-614EA 12 USD InStock
77244-614 77244-616 77244-618 77244-620
Fmoc-(Nd-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl)-D-ornithine 97%
Fmoc-(Nd-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl)-D-ornithine
Precaución: for Research Use Only


Formula: C₃₀H₃₄N₂O₆
MW: 518.62 g/mol
MDL Number: MFCD02094127
CAS Number: 1419640-31-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty