Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fast Blue BB base 98%

Supplier: AMBEED, INC
Danger

A879635-1G A879635-250MG A879635-5G
77294-106EA 13 USD InStock
77294-106 77294-108 77294-110
Fast Blue BB base 98%
Fast Blue BB base
Precaución: for Research Use Only


Formula: C₁₇H₂₀N₂O₃
MW: 300.36 g/mol
MDL Number: MFCD00009091
CAS Number: 120-00-3
UN: 3259
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty