Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-(4-((3-((2-Methylbenzo[d]thiazol-6-yl)amino)-3-oxopropyl)amino)-4-oxobutyl)benzyl)malonic acid 97%

Supplier: AMBEED, INC
Warning

A879979-10MG A879979-1MG A879979-50MG A879979-5MG
77244-628EA 60 USD InStock
77244-628 77244-630 77244-632 77244-634
2-(4-(4-((3-((2-Methylbenzo[d]thiazol-6-yl)amino)-3-oxopropyl)amino)-4-oxobutyl)benzyl)malonic acid 97%
2-(4-(4-((3-((2-Methylbenzo[d]thiazol-6-yl)amino)-3-oxopropyl)amino)-4-oxobutyl)benzyl)malonic acid


Formula: C₂₅H₂₇N₃O₆S
MDL Number: MFCD29913759
CAS Number: 1370290-34-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty