Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-3-furanthiol 95%

Supplier: AMBEED, INC
Danger

A884716-250MG A884716-25G A884716-5G A884716-1G
77281-838EA 7 USD InStock
77281-838 77281-840 77281-842 77749-690
2-Methyl-3-furanthiol 95%
2-Methyl-3-furanthiol
Precaución: for Research Use Only


Formula: C₅H₆OS
MW: 114.16 g/mol
MDL Number: MFCD00010280
CAS Number: 28588-74-1
UN: 1228
ADR: 3,6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty