Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-((3,5-Bis(trifluoromethyl)phenyl)amino)-4-(((1R,2R)-2-(dimethylamino)cyclohexyl)amino)cyclobut-3-ene-1,2-dione 98%

Supplier: AMBEED, INC
Warning

A891214-1G A891214-250MG A891214-100MG
77256-494EA 294.99 USD InStock
77256-494 77256-496 77747-651
3-((3,5-Bis(trifluoromethyl)phenyl)amino)-4-(((1R,2R)-2-(dimethylamino)cyclohexyl)amino)cyclobut-3-ene-1,2-dione 98%
3-((3,5-Bis(trifluoromethyl)phenyl)amino)-4-(((1R,2R)-2-(dimethylamino)cyclohexyl)amino)cyclobut-3-ene-1,2-dione
Precaución: for Research Use Only


Formula: C₂₀H₂₁F₆N₃O₂
MW: 449.39 g/mol
CAS Number: 1211565-07-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty