Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-([1,4'-Bipiperidin]-1'-ylmethyl)-7-bromo-N-(1-phenylcyclopropyl)-2-(3-(trifluoromethyl)phenyl)quinoline-4-carboxamide 98%

Supplier: AMBEED, INC
Danger

A892019-10MG A892019-1MG A892019-5MG
77268-760EA 109 USD InStock
77268-760 77268-762 77268-764
3-([1,4'-Bipiperidin]-1'-ylmethyl)-7-bromo-N-(1-phenylcyclopropyl)-2-(3-(trifluoromethyl)phenyl)quinoline-4-carboxamide 98%
3-([1,4'-Bipiperidin]-1'-ylmethyl)-7-bromo-N-(1-phenylcyclopropyl)-2-(3-(trifluoromethyl)phenyl)quinoline-4-carboxamide
Precaución: for Research Use Only


Formula: C₃₇H₃₈BrF₃N₄O
MDL Number: MFCD28009510
CAS Number: 1336960-13-4
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty