Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2'-Bipyridyl-1,1'-dioxide 98%

Supplier: AMBEED, INC
Warning

A896050-100G A896050-1G A896050-25G A896050-5G
77256-514EA 342 USD InStock
77256-514 77256-516 77256-518 77256-520
2,2'-Bipyridyl-1,1'-dioxide 98%
2,2'-Bipyridyl-1,1'-dioxide
Precaución: for Research Use Only


Formula: C₁₀H₈N₂O₂
MDL Number: MFCD00065169
CAS Number: 7275-43-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty