Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-tert-Butyl (2-oxopiperidin-4-yl)carbamate 97%

Supplier: AMBEED, INC
Warning

A899742-100MG A899742-250MG
77268-810EA 63 USD InStock
77268-810 77268-812
(S)-tert-Butyl (2-oxopiperidin-4-yl)carbamate 97%
(S)-tert-Butyl (2-oxopiperidin-4-yl)carbamate
Precaución: for Research Use Only


Formula: C₁₀H₁₈N₂O₃
MDL Number: MFCD30730061
CAS Number: 1055049-80-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty