Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Hydroxymethyl)-5-methylisoxazole 98%

Supplier: AMBEED, INC
Warning

A901742-1G A901742-250MG A901742-5G
77294-196EA 34 USD InStock
77294-196 77294-198 77294-200
3-(Hydroxymethyl)-5-methylisoxazole 98%
3-(Hydroxymethyl)-5-methylisoxazole
Precaución: for Research Use Only


Formula: C₅H₇NO₂
MDL Number: MFCD00085129
CAS Number: 35166-33-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty