Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dibromo-4-chloroaniline 97%

Supplier: AMBEED, INC
Warning

A901977-100G A901977-5G A901977-25G
77294-202EA 53 USD InStock
77294-202 77294-204 77749-712
2,6-Dibromo-4-chloroaniline 97%
2,6-Dibromo-4-chloroaniline
Precaución: for Research Use Only


Formula: C₆H₄Br₂ClN
MDL Number: MFCD00051750
CAS Number: 874-17-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty