Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dibromo-3-phenylpropionic acid 98%

Supplier: AMBEED, INC
Warning

A904045-10G A904045-1G A904045-25G A904045-5G
77256-532EA 37 USD InStock
77256-532 77256-534 77256-536 77256-538
2,3-Dibromo-3-phenylpropionic acid 98%
2,3-Dibromo-3-phenylpropionic acid
Precaución: for Research Use Only


Formula: C₉H₈Br₂O₂
MDL Number: MFCD00004208
CAS Number: 6286-30-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty