Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-3,4-dimethoxybenzaldehyde 97%

Supplier: AMBEED, INC
Warning

A904947-100MG A904947-250MG A904947-5G
77268-818EA 50 USD InStock
77268-818 77268-820 77268-822
2-Fluoro-3,4-dimethoxybenzaldehyde 97%
2-Fluoro-3,4-dimethoxybenzaldehyde
Precaución: for Research Use Only


Formula: C₉H₉FO₃
MDL Number: MFCD03411462
CAS Number: 37686-68-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty