Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trifluoromethyl)benzylamine 95%

Supplier: AMBEED, INC
Danger

A905139-10G A905139-25G A905139-5G
77244-736EA 19 USD InStock
77244-736 77244-738 77244-740 77244-742
2-(Trifluoromethyl)benzylamine 95%
2-(Trifluoromethyl)benzylamine
Precaución: for Research Use Only


Formula: C₈H₈F₃N
MW: 175.15
Boiling Pt: 108
MDL Number: MFCD00010297
CAS Number: 3048-01-9
UN: 2735
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty