Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-methyl-3-pyridineboronic acid 97%

Supplier: AMBEED, INC
Warning

A909049-1G A909049-5G
77268-836EA 28 USD InStock
77268-836 77268-838 77268-840
2-Chloro-5-methyl-3-pyridineboronic acid 97%
2-Chloro-5-methyl-3-pyridineboronic acid
Precaución: for Research Use Only


Formula: C₆H₇BClNO₂
MDL Number: MFCD04112563
CAS Number: 913835-86-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty