Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-chloro-3-nitrobenzotrifluoride 98%

Supplier: AMBEED, INC
Warning

A911205-1G A911205-250MG A911205-5G
77256-560EA 14 USD InStock
77256-560 77256-562 77256-564
2-Amino-5-chloro-3-nitrobenzotrifluoride 98%
2-Amino-5-chloro-3-nitrobenzotrifluoride
Precaución: for Research Use Only


Formula: C₇H₄ClF₃N₂O₂
MDL Number: MFCD03094262
CAS Number: 62924-50-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty