Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methylglutaric anhydride 96%

Supplier: AMBEED, INC
Warning

A918833-100G A918833-1G A918833-25G A918833-5G
77281-928EA 188 USD InStock
77281-928 77281-930 77281-932 77281-934
3-Methylglutaric anhydride 96%
3-Methylglutaric anhydride
Precaución: for Research Use Only


Formula: C₆H₈O₃
MW: 128.1259 g/mol
MDL Number: MFCD00006683
CAS Number: 4166-53-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty