Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Phenylazophenol 95% (by HPLC)

Supplier: AMBEED, INC
Warning

A919177-100G A919177-10G A919177-25G A919177-5G
77294-228EA 105 USD InStock
77294-228 77294-230 77294-232 77294-234
4-Phenylazophenol 95% (by HPLC)
4-Phenylazophenol
Precaución: for Research Use Only


Formula: C₁₂H₁₀N₂O
MDL Number: MFCD00002330
CAS Number: 1689-82-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty