Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-phenyl-1,2,4-oxadiazole 95+%

Supplier: AMBEED, INC
Warning

A925757-100MG A925757-1G A925757-250MG
77281-958EA 50 USD InStock
77281-958 77281-960 77281-962
3-Bromo-5-phenyl-1,2,4-oxadiazole 95+%
3-Bromo-5-phenyl-1,2,4-oxadiazole
Precaución: for Research Use Only


Formula: C₈H₅BrN₂O
MW: 225.05 g/mol
MDL Number: MFCD06795878
CAS Number: 23432-94-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty