Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium (3,6-dihydro-2H-pyran-4-yl)trifluoroborate 97%

Supplier: AMBEED, INC
Danger

A929168-1G A929168-250MG A929168-5G
77244-818EA 219 USD InStock
77244-818 77244-820 77244-822
Potassium (3,6-dihydro-2H-pyran-4-yl)trifluoroborate 97%
Potassium (3,6-dihydro-2H-pyran-4-yl)trifluoroborate
Precaución: for Research Use Only


Formula: C₅H₇BF₃KO
MDL Number: MFCD12827761
CAS Number: 1612893-03-4
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty