Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-2-isopropoxyaniline hydrochloride 98%

Supplier: AMBEED, INC
Warning

A931780-100MG A931780-1G A931780-250MG
77294-286EA 6 USD InStock
77294-286 77294-288 77294-290
4-Fluoro-2-isopropoxyaniline hydrochloride 98%
4-Fluoro-2-isopropoxyaniline hydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₃ClFNO
MDL Number: MFCD01632194
CAS Number: 380430-47-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty