Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2,6-difluorobenzyl bromide 95%

Supplier: AMBEED, INC
Danger

A937952-100MG A937952-1G A937952-250MG
77294-304EA 22 USD InStock
77294-304 77294-306 77294-308
4-Bromo-2,6-difluorobenzyl bromide 95%
4-Bromo-2,6-difluorobenzyl bromide
Precaución: for Research Use Only


Formula: C₇H₄Br₂F₂
MDL Number: MFCD03094462
CAS Number: 162744-60-7
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty