Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(5-Methoxy-5-oxopentyl)triphenylphosphonium bromide 95%

Supplier: AMBEED, INC
Warning

A940851-100G A940851-1G A940851-25G A940851-5G
77256-724EA 395 USD InStock
77256-724 77256-726 77256-728 77256-730
(5-Methoxy-5-oxopentyl)triphenylphosphonium bromide 95%
(5-Methoxy-5-oxopentyl)triphenylphosphonium bromide
Precaución: for Research Use Only


Formula: C₂₄H₂₆BrO₂P
MDL Number: MFCD28347730
CAS Number: 79837-79-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty