Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ω,ω-Diethoxyacetophenone 98%

Supplier: AMBEED, INC
Warning

A946980-1000G A946980-100G A946980-25G A946980-500G
77256-766EA 186 USD InStock
77256-766 77256-768 77256-770 77256-772
ω,ω-Diethoxyacetophenone 98%
ω,ω-Diethoxyacetophenone
Precaución: for Research Use Only


Formula: C₁₂H₁₆O₃
MDL Number: MFCD00009659
CAS Number: 6175-45-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty