Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Nitro-4-(trifluoromethyl)phenylacetonitrile 97%

Supplier: AMBEED, INC
Warning

A951624-1G A951624-250MG A951624-5G
77282-042EA 23 USD InStock
77282-042 77282-044 77282-046
2-Nitro-4-(trifluoromethyl)phenylacetonitrile 97%
2-Nitro-4-(trifluoromethyl)phenylacetonitrile


Formula: C₉H₅F₃N₂O₂
MDL Number: MFCD00045621
CAS Number: 13544-06-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty