Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Ethylcyclopentanol 98%

Supplier: AMBEED, INC
Danger

A952628-25G A952628-5G
77244-908EA 52 USD InStock
77244-908 77244-910
1-Ethylcyclopentanol 98%
1-Ethylcyclopentanol
Precaución: for Research Use Only


Formula: C₇H₁₄O
MDL Number: MFCD00019281
CAS Number: 1462-96-0
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty