Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-1,3,5-benzenetricarbaldehyde 95%

Supplier: AMBEED, INC
Warning

A954938-100MG A954938-1G A954938-250MG A954938-10G
77269-062EA 13 USD InStock
77269-062 77269-064 77269-066 77749-784
2-Hydroxy-1,3,5-benzenetricarbaldehyde 95%
2-Hydroxy-1,3,5-benzenetricarbaldehyde
Precaución: for Research Use Only


Formula: C₉H₆O₄
MDL Number: MFCD01314207
CAS Number: 81502-74-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty