Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Ethoxyacrylonitrile (cis- and trans- mixture) 95%

Supplier: AMBEED, INC
Warning

A956940-100G A956940-1G A956940-25G A956940-5G
77282-048EA 125 USD InStock
77282-048 77282-050 77282-052 77282-054
3-Ethoxyacrylonitrile (cis- and trans- mixture) 95%
3-Ethoxyacrylonitrile (cis- and trans- mixture)
Precaución: for Research Use Only


Formula: C₅H₇NO
MDL Number: MFCD00010193
CAS Number: 61310-53-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty