Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-tert-Butyl 2-amino-2-phenylacetate hydrochloride 97%

Supplier: AMBEED, INC
Warning

A957591-1G A957591-25G A957591-5G A957591-100G
77256-824EA 7 USD InStock
77256-824 77256-826 77256-828 77747-713
(R)-tert-Butyl 2-amino-2-phenylacetate hydrochloride 97%
(R)-tert-Butyl 2-amino-2-phenylacetate hydrochloride
Precaución: for Research Use Only


Formula: C₁₂H₁₈ClNO₂
MW: 243.72 g/mol
MDL Number: MFCD03093480
CAS Number: 256478-95-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty