Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Nitroethanol 97%

Supplier: AMBEED, INC
Danger

A957769-100G A957769-10G A957769-1G A957769-25G A957769-5G
77282-056EA 167 USD InStock
77282-056 77282-058 77282-060 77282-062 77282-064
2-Nitroethanol 97%
2-Nitroethanol
Precaución: for Research Use Only


Formula: C₂H₅NO₃
MDL Number: MFCD00007405
CAS Number: 625-48-9
UN: 1993
ADR: 3,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty