Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Methoxyphenoxy)ethylamine 98%

Supplier: AMBEED, INC
Danger

A957783-10G A957783-25G
77256-830EA 7 USD InStock
77256-830 77256-832
2-(2-Methoxyphenoxy)ethylamine 98%
2-(2-Methoxyphenoxy)ethylamine
Precaución: for Research Use Only


Formula: C₉H₁₃NO₂
MDL Number: MFCD00235185
CAS Number: 1836-62-0
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty