Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Pyrrolidin-2-yl)phenol hydrochloride 97%

Supplier: AMBEED, INC
Warning

A957785-100MG A957785-1G A957785-250MG
77256-834EA 86 USD InStock
77256-834 77256-836 77256-838
3-(Pyrrolidin-2-yl)phenol hydrochloride 97%
3-(Pyrrolidin-2-yl)phenol hydrochloride
Precaución: for Research Use Only


Formula: C₈H₈F₃N₃OS
MW: 199.68 g/mol
MDL Number: MFCD30471573
CAS Number: 1894060-74-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty