Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

AZD-5582 ≥98%

Supplier: AMBEED, INC
Warning

A961946-10MG A961946-1MG A961946-25MG A961946-50MG A961946-5MG
77244-936EA 106 USD InStock
77244-936 77244-938 77244-940 77244-942 77244-944
AZD-5582 ≥98%
AZD-5582
Precaución: for Research Use Only


Formula: C₅₈H₇₈N₈O₈
MW: 1015.29 g/mol
MDL Number: MFCD28411397
CAS Number: 1258392-53-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty