Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

GNE-317 98%

Supplier: AMBEED, INC
Warning

A964728-10MG A964728-1MG A964728-50MG A964728-5MG
77244-948EA 100 USD InStock
77244-948 77244-950 77244-952 77244-954
GNE-317 98%
GNE-317
Precaución: for Research Use Only


Formula: C₁₉H₂₂N₆O₃S
MDL Number: MFCD28900677
CAS Number: 1394076-92-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty