Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[2,2'-Bipyridine]-5,5'-dicarboxaldehyde 97%

Supplier: AMBEED, INC
Warning

A966154-100MG A966154-1G A966154-250MG A966154-5G
77256-878EA 14 USD InStock
77256-878 77256-880 77256-882 77256-884
[2,2'-Bipyridine]-5,5'-dicarboxaldehyde 97%
[2,2'-Bipyridine]-5,5'-dicarboxaldehyde
Precaución: for Research Use Only


Formula: C₁₂H₈N₂O₂
MDL Number: MFCD03093215
CAS Number: 135822-72-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty