Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methanesulfonato(2-dicyclohexylphosphino-2',6'-di-i-propoxy-1,1'-biphenyl)(2'-methylamino-1,1'-biphenyl-2-yl)palladium(II) 98%

Supplier: AMBEED, INC
Warning

A971331-100MG A971331-1G A971331-250MG A971331-5G A971331-25G
77256-900EA 10 USD InStock
77256-900 77256-902 77256-904 77256-906 77747-727
Methanesulfonato(2-dicyclohexylphosphino-2',6'-di-i-propoxy-1,1'-biphenyl)(2'-methylamino-1,1'-biphenyl-2-yl)palladium(II) 98%
Methanesulfonato(2-dicyclohexylphosphino-2',6'-di-i-propoxy-1,1'-biphenyl)(2'-methylamino-1,1'-biphenyl-2-yl)palladium(II)
Precaución: for Research Use Only


Formula: C₄₄H₅₈NO₅PPdS
MW: 850.39 g/mol
MDL Number: MFCD28411659
CAS Number: 1599466-85-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty