Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-2,2,4-Trimethyl-1,3-pentanediol 97%

Supplier: AMBEED, INC
Warning

A981137-1000G A981137-100G A981137-25G A981137-500G
77256-960EA 41 USD InStock
77256-960 77256-962 77256-964 77256-966
(±)-2,2,4-Trimethyl-1,3-pentanediol 97%
(±)-2,2,4-Trimethyl-1,3-pentanediol
Precaución: for Research Use Only


Formula: C₈H₁₈O₂
MW: 146.23 g/mol
Boiling Pt: 228.9
Melting Pt: 51
MDL Number: MFCD00004681
CAS Number: 144-19-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty