Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Guanine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A989116-25G A989116-5G
77256-988EA 10.3 USD InStock
77256-988 77256-990 77256-992
Guanine hydrochloride 98%
Guanine hydrochloride
Precaución: for Research Use Only


Formula: C₅H₆ClN₅O
MDL Number: MFCD00213670
CAS Number: 635-39-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty